4-Hydroxy-4'-nitrostilbene 98%
Leverantör: Thermo Scientific
|
Varning
|
B20913.03
B20913.06
B20913.03EA
955
SEK
B20913.03
B20913.06
4-Hydroxy-4'-nitrostilbene 98%
4-Hydroxy-4'-nitrostilbene
Purity: 98%
CAS No.: 19221-08-0
Molecular Formula: C14H11NO3
Formula Weight: 241.25
Melting Point: 205-209°
Beilstein Reference: 2505854
MDL No.: MFCD00017044
Formel:
C₁₄H₁₁NO₃ MW: 241,25 g/mol Smältpunkt: 205…209 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00017044 CAS nummer: 19221-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...