2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Hydrazinyl-4,5-dihydro-1H-imidazole hydrobromide, 2-Hydrazino-2-imidazoline Hydrobromide, 2-Hydrazino-4,5-dihydroimidazole Hydrobromide
B20386.09 B20386.88
B20386.09EA 5400 SEK
B20386.09 B20386.88
2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide 98%
2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide
Purity: 98% CAS No.: 55959-84-7 Molecular Formula: C3H8N4.HBr Formula Weight: 181.04 Melting Point: 181-185° MDL No.: MFCD00012694
Formel: C₃H₉BrN₄
MW: 181,04 g/mol
Smältpunkt: 181…185 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012694
CAS nummer: 55959-84-7
EINECS: 259-921-3