3-Hydroxy-1-adamantane carboxylic acid 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Hydroxyadamantane-1-carboxylic acid, 3-Hydroxy-1-adamantanecarboxylic acid
B20377.03 B20377.06
B20377.03EA 1675 SEK
B20377.03 B20377.06
3-Hydroxy-1-adamantane carboxylic acid 97%
3-Hydroxy-1-adamantane carboxylic acid
Purity: 98% CAS No.: 42711-75-1 Molecular Formula: C11H16O3 Formula Weight: 196.25 Melting Point: 203-207° MDL No.: MFCD00193929
Formel: C₁₁H₁₆O₃
MW: 196.25 g/mol
Smältpunkt: 203…207 °C
Storage Temperature: Omgivande
MDL Number: MFCD00193929
CAS nummer: 42711-75-1