Ethyl-2-oxo-4-phenylbutyrate ≥90%
Leverantör: Thermo Scientific
Synonyms: Benzylpyruvic Acid Ethyl Ester, Ethyl Benzylpyruvate, 2-Oxo-4-phenylbutyric Acid Ethyl Ester
B20148.09 B20148.18
B20148.09EA 1750 SEK
B20148.09 B20148.18
Ethyl-2-oxo-4-phenylbutyrate ≥90%
Ethyl-2-oxo-4-phenylbutyrate
Purity: 90+% CAS No.: 64920-29-2 Molecular Formula: C12H14O3 Formula Weight: 206.24 Flash Point: 110°(230°F) Refractive Index: 1.5020 Boiling Point: 131-133°/2mm Beilstein Reference: 2725083 MDL No.: MFCD00037533
Formel: C₁₂H₁₄O₃
MW: 206,24 g/mol
Kokpunkt: 131…133 °C (2 mmHg)
Densitet: 1,09
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00037533
CAS nummer: 64920-29-2
EINECS: 265-276-9