Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride 97%
Leverantör: Thermo Scientific
Varning
A19632.14 A19632.22
A19632.14EA 1300 SEK
A19632.14 A19632.22
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride 97%
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride
Purity: 97% CAS No.: 1719-83-1 Molecular Formula: C12H8O6 Formula Weight: 248.19 Melting Point: ca 360° Beilstein Reference: 294213 MDL No.: MFCD00082228
Formel: C₁₂H₈O₆
MW: 248,19 g/mol
Smältpunkt: ∼ 360 °C
Storage Temperature: Omgivande
MDL Number: MFCD00082228
CAS nummer: 1719-83-1
EINECS: 217-009-2