Ethyl-2-methyl-4-pentenoate 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Ethyl 2-methylpent-4-enoate, 2-Methyl-4-pentenoic Acid Ethyl Ester
A19526.14 A19526.22
A19526.14EA 1950 SEK
A19526.14 A19526.22
Ethyl-2-methyl-4-pentenoate 98%
Ethyl-2-methyl-4-pentenoate
Purity: 98% CAS No.: 53399-81-8 Molecular Formula: C8H14O2 Formula Weight: 142.20 Flash Point: 40°(104°F) Refractive Index: 1.4160 Boiling Point: 153-155° Beilstein Reference: 1753406 UN No.: UN3272 MDL No.: MFCD00010235
Formel: C₈H₁₄O₂
MW: 142,20 g/mol
Kokpunkt: 153…155 °C
Densitet: 0,880
Flampunkt: 40 °C (104 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00010235
CAS nummer: 53399-81-8
EINECS: 258-520-0
ADR: 3,III