(4-tert-Butylphenoxy)acetyl chloride 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-(4-(tert-Butyl)phenoxy)acetyl chloride
A19408.09 A19408.18
A19408.09EA 1325 SEK
A19408.09 A19408.18
(4-tert-Butylphenoxy)acetyl chloride 98%
(4-tert-Butylphenoxy)acetyl chloride
Purity: 98% CAS No.: 90734-55-7 Molecular Formula: C12H15O2Cl Formula Weight: 226.71 Refractive Index: 1.5165 Boiling Point: 130-132°/5mm UN No.: UN3265 MDL No.: MFCD00798568
Formel: C₁₂H₁₅ClO₂
MW: 226,71 g/mol
Kokpunkt: 130…132 °C (5 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD00798568
CAS nummer: 90734-55-7
ADR: 8,II