4,4'-Methylenebis(2,6-di-tert-butylphenol) 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Bis(3,5-di-tert-butyl-4-hydroxyphenyl)methane
A18910.18 A18910.22
A18910.18EA 570 SEK
A18910.18 A18910.22
4,4'-Methylenebis(2,6-di-tert-butylphenol) 98%
4,4'-Methylenebis(2,6-di-tert-butylphenol)
Purity: 98% CAS No.: 118-82-1 Molecular Formula: C29H44O2 Formula Weight: 424.67 Melting Point: 154-158° Boiling Point: 289°/40mm Beilstein Reference: 1916919 MDL No.: MFCD00008822
Formel: C₂₉H₄₄O₂
MW: 424,67 g/mol
Kokpunkt: 289 °C (40 mmHg)
Smältpunkt: 154…158 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008822
CAS nummer: 118-82-1
EINECS: 204-279-1