Lithium formate monohydrate 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Formic Acid Lithium Salt, Lithium formate hydrate
A17432.18 A17432.30
A17432.18EA 690 SEK
A17432.18 A17432.30
Lithium formate monohydrate 98%
Lithium formate monohydrate
Purity: 98% CAS No.: 6108-23-2 Molecular Formula: LiOOCH.xH2O Formula Weight: 51.96(anhy) Odour: Mild Melting Point: 94° dec. MDL No.: MFCD00050823
Formel: HCO₂Li·H₂O
MW: 51.96 g/mol (anhy)
Smältpunkt: 94 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00050823
CAS nummer: 6108-23-2
EINECS: 209-133-0