Ethyl-3-aminopropanoate hydrochloride 98%
Leverantör: Thermo Scientific
Synonyms: beta-Alanine ethyl ester hydrochloride, Ethyl 3-Aminopropionate Hydrochloride, 3-Aminopropionic Acid Ethyl Ester Hydrochloride, Ethyl 3-aminopropanoate hydrochloride, H-β-Ala-OEt·HCl, Ethyl 3-aminopropionate HCl
A17277.09 A17277.18 A17277.30
A17277.09EA 1250 SEK
A17277.09 A17277.18 A17277.30
Ethyl-3-aminopropanoate hydrochloride 98%
Ethyl-3-aminopropanoate hydrochloride
Purity: 98% CAS No.: 4244-84-2 Molecular Formula: H2NCH2CH2CO2CH2CH3.HCl Formula Weight: 153.61 Melting Point: 67-72° Beilstein Reference: 3559095 MDL No.: MFCD00012909
Formel: C₅H₁₁NO₂·HCl
MW: 153,6 g/mol
Smältpunkt: 67…72 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012909
CAS nummer: 4244-84-2
EINECS: 224-203-0