1-Acetamidoadamantane 98%
Leverantör: Thermo Scientific
Varning
Synonyms: N-1-Adamantylacetamide, N-Acetyl-1-adamantanamine
A17224.09 A17224.18
A17224.09EA 1025 SEK
A17224.09 A17224.18
1-Acetamidoadamantane 98%
1-Acetamidoadamantane
Purity: 98% CAS No.: 880-52-4 Molecular Formula: C12H19NO Formula Weight: 193.29 Melting Point: 146-149° MDL No.: MFCD00074730
Formel: C₁₂H₁₉NO
MW: 193,29 g/mol
Smältpunkt: 146…149 °C
Storage Temperature: Omgivande
MDL Number: MFCD00074730
CAS nummer: 880-52-4
EINECS: 212-914-9