Tris(ethylenediamine)cobalt(III) chloride trihydrate
Leverantör: Thermo Scientific
Varning
A17211.09 A17211.18
A17211.09EA 1375 SEK
A17211.09 A17211.18
Tris(ethylenediamine)cobalt(III) chloride trihydrate
Tris(ethylenediamine)cobalt(III) chloride trihydrate
CAS No.: 14883-80-8 Molecular Formula: C6H24Cl3CoN6.3H2O Formula Weight: 399.64 (345.59anhy) MDL No.: MFCD00211318
Formel: C₆H₂₄Cl₃CoN₆·3H₂O
MW: 399,64 g/mol (345,59 anhy)
Storage Temperature: Omgivande
MDL Number: MFCD00211318
CAS nummer: 14883-80-8