Menthofuran 95%
Leverantör: Thermo Scientific
Varning
|
A17192.03
A17192.06
A17192.03EA
1175
SEK
A17192.03
A17192.06
Menthofuran 95%
Menthofuran
Purity: 95%
CAS No.: 494-90-6
Molecular Formula: C10H14O
Formula Weight: 150.22
Flash Point: 75°(167°F)
Refractive Index: 1.4845
Boiling Point: 86-88°/15mm
MDL No.: MFCD00041851
Formel:
C₁₀H₁₄O MW: 150,221 g/mol Kokpunkt: 86…88 °C (15 mmHg) Densitet: 0,972 Flampunkt: 75 °C (167 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00041851 CAS nummer: 494-90-6 EINECS: 207-795-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...