cis-1,2-Bis(diphenylphosphino)ethylene 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (Z)-1,2-Bis(diphenylphosphino)ethene
A16859.03 A16859.06
A16859.03EA 2050 SEK
A16859.03 A16859.06
cis-1,2-Bis(diphenylphosphino)ethylene 97%
cis-1,2-Bis(diphenylphosphino)ethylene
Purity: 97% CAS No.: 983-80-2 Molecular Formula: C26H22P2 Formula Weight: 396.41 Melting Point: 122-126° Beilstein Reference: 757792 MDL No.: MFCD00003046
Formel: C₂₆H₂₂P₂
Smältpunkt: 122…126 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003046
CAS nummer: 983-80-2
EINECS: 213-569-7