1,6-Anhydro-β-D-glucose 99%
Leverantör: Thermo Scientific
Synonyms: 1,6-Anhydro-β-D-glucopyranose, (1R,2S,3S,4R,5R)-6,8-Dioxabicyclo[3.2.1]octane-2,3,4-triol, 1,6-Anhydro-β-D-glucose, (-)-1,6-Anhydro-β-D-glucopyranose, Levoglucosan, (-)-1,6-Anhydro-ß-D-glucopyranose
A16724.01 A16724.03
A16724.01EA 690 SEK
A16724.01 A16724.03
1,6-Anhydro-β-D-glucose 99%
1,6-Anhydro-beta-D-glucose
Purity: 99% CAS No.: 498-07-7 Molecular Formula: C6H10O5 Formula Weight: 162.14 Melting Point: 180-184° Beilstein Reference: 80998 MDL No.: MFCD00063248
Formel: C₆H₁₀O₅
MW: 162,14 g/mol
Smältpunkt: 180…184 °C
Storage Temperature: Omgivande
MDL Number: MFCD00063248
CAS nummer: 498-07-7
EINECS: 207-855-0