2-(1-Naphthyl)-5-phenyloxazole ≥99% laser grade, for scintillation
Leverantör: Thermo Scientific
Synonyms: ANPO
A16679.03 A16679.06
A16679.03EA 900 SEK
A16679.03 A16679.06
2-(1-Naphthyl)-5-phenyloxazole ≥99% laser grade, for scintillation
2-(1-Naphthyl)-5-phenyloxazole
Purity: 99+% CAS No.: 846-63-9 Molecular Formula: C19H13NO Formula Weight: 271.32 Melting Point: 104-105° Beilstein Reference: 210666 MDL No.: MFCD00019714
Formel: C₁₉H₁₃NO
MW: 271,32 g/mol
Smältpunkt: 104…105 °C
Storage Temperature: Omgivande
MDL Number: MFCD00019714
CAS nummer: 846-63-9
EINECS: 212-689-7