2,4,6-Trimethylstyrene 95% stabilised
Leverantör: Thermo Fisher Scientific
|
Varning
|
Stabilization:
med 4-tert-Butylpyrokatekol 500 ppm
Stab. with 500ppm 4-tert-butylcatechol
Purity: 95%
CAS No.: 769-25-5
Molecular Formula: C11H14
Formula Weight: 146.23
Storage Temperature: 2°C to 8°C
Flash Point: 75°(167°F)
Refractive Index: 1.5320
Boiling Point: 208-209°
Beilstein Reference: 1904365
MDL No.: MFCD00008613
Formel:
C₁₁H₁₄ Kokpunkt: 208…209 °C Densitet: 0,908 Flampunkt: 75 °C (167 °F) Storage Temperature: Kylskåp |
MDL Number:
MFCD00008613 CAS nummer: 769-25-5 EINECS: 212-205-4 |