Ethoxyacetylene 50% (w/w) i hexan
Leverantör: Thermo Scientific
Fara
Synonyms: ethynoxyethane, Ethyl ethynyl ether, ethoxy-ethyne
A14245.03 A14245.06 A14245.14
A14245.03EA 1325 SEK
A14245.03 A14245.06 A14245.14
Ethoxyacetylene 50% (w/w) i hexan
Ethoxyacetylene
CAS No.: 927-80-0 Molecular Formula: C4H6O Formula Weight: 70.09 Storage Temperature: 2°C to 8°C Flash Point: -28°(-18°F) Refractive Index: 1.3800 Odour: Recognizable Boiling Point: 55-56° Beilstein Reference: 741882 UN No.: UN1993 MDL No.: MFCD00009247
Formel: C₄H₆O
MW: 70,091 g/mol
Kokpunkt: 55…56 °C
Densitet: 0,718
Flampunkt: –28 °C (–18 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00009247
CAS nummer: 927-80-0
EINECS: 213-164-5
UN: 1993
ADR: 3,II