Monoacetin mixture of isomers cont. varying amounts of diacetate, tech.
Leverantör: Thermo Scientific
Synonyms: Glycerol Monoacetate, 1,2,3-Propanetriol, monoacetate
A14025.30 A14025.0B
A14025.30EA 2850 SEK
A14025.30 A14025.0B
Monoacetin mixture of isomers cont. varying amounts of diacetate, tech.
Monoacetin
CAS No.: 26446-35-5 Molecular Formula: C5H10O4 Formula Weight: 134.13 Flash Point: 145°(293°F) Refractive Index: 1.4500 Boiling Point: 258° MDL No.: MFCD00036185
Formel: C₅H₁₀O₄
MW: 134,13 g/mol
Kokpunkt: 258 °C
Densitet: 1,21
Flampunkt: 145 °C (293 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00036185
CAS nummer: 26446-35-5
EINECS: 247-704-6
Merck Index: 14,6242