Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate 97%
Leverantör: Thermo Scientific
Synonyms: Ethyl 3-(trifluoromethyl)pyrazole-4-carboxylate, 3-(Trifluoromethyl)pyrazole-4-carboxylic Acid Ethyl Ester, Ethyl 3-trifluoromethylpyrazole-4-carboxylate
A13609.06 A13609.14
A13609.06EA 2850 SEK
A13609.06 A13609.14
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate 97%
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate
Purity: 97% CAS No.: 155377-19-8 Molecular Formula: C7H7F3N2O2 Formula Weight: 208.14 Melting Point: 142-146° MDL No.: MFCD00052083
Formel: C₇H₇F₃N₂O₂
MW: 208,14 g/mol
Smältpunkt: 142…146 °C
Storage Temperature: Omgivande
MDL Number: MFCD00052083
CAS nummer: 155377-19-8