Methyl-4-(methylamino)benzoate 98%
Leverantör: Thermo Fisher Scientific
Synonyms: Methyl 4-(methylamino)benzoate
A13460.06 A13460.14
A13460.06EA 1575 SEK
A13460.06 A13460.14
Methyl-4-(methylamino)benzoate 98%
Methyl-4-(methylamino)benzoate
Purity: 98% CAS No.: 18358-63-9 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Melting Point: 92-97° Beilstein Reference: 2803327 MDL No.: MFCD00017198
Formel: C₉H₁₁NO₂
MW: 165.19 g/mol
Smältpunkt: 92…97 °C
Storage Temperature: Omgivande
MDL Number: MFCD00017198
CAS nummer: 18358-63-9