N-Hydroxy-5-norbornene-2,3-dicarboximide 97%
Leverantör: Thermo Scientific
Varning
Synonyms: endo-N-Hydroxybicyclo[2.2.1]hept-5-ene-2,3-dicarboximide
A13205.09 A13205.18 A13205.30
A13205.09EA 765 SEK
A13205.09 A13205.18 A13205.30
N-Hydroxy-5-norbornene-2,3-dicarboximide 97%
N-Hydroxy-5-norbornene-2,3-dicarboximide
Purity: 97% CAS No.: 21715-90-2 Molecular Formula: C9H9NO3 Formula Weight: 179.18 Melting Point: 166-170° Beilstein Reference: 13540 MDL No.: MFCD00065691
Formel: C₉H₉NO₃
MW: 179,18 g/mol
Smältpunkt: 166…170 °C
Storage Temperature: Omgivande
MDL Number: MFCD00167583
CAS nummer: 21715-90-2
EINECS: 244-538-6