1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,2',2'',2'''-(((Ethane-1,2-diylbis(oxy))bis(2,1-phenylene))bis(azanetriyl))tetraacetic acid, BAPTA
A13190.03 A13190.06 A13190.14
A13190.03EA 3150 SEK
A13190.03 A13190.06 A13190.14
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid
Purity: 97% CAS No.: 85233-19-8 Molecular Formula: C22H24N2O10 Formula Weight: 476.44 Storage Temperature: 2°C to 8°C Melting Point: ca 174° dec. MDL No.: MFCD00036255
Formel: C₂₂H₂₄N₂O₁₀
MW: 476,44 g/mol
Smältpunkt: ∼ 174 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00036255
CAS nummer: 85233-19-8
Merck Index: 14,957