Ethyl cyano(hydroxyimino)acetate 97%
Leverantör: Thermo Scientific
Fara
Synonyms: Ethyl 2-cyano-2-(hydroxyimino)acetate, Ethyl Cyanoglyoxalate Oxime, Cyano(hydroxyimino)acetic Acid Ethyl Ester, Oxyma
A12691.06 A12691.14 A12691.22
A12691.06EA 670 SEK
A12691.06 A12691.14 A12691.22
Ethyl cyano(hydroxyimino)acetate 97%
Ethyl cyano(hydroxyimino)acetate
Purity: 97% CAS No.: 3849-21-6 Molecular Formula: C5H6N2O3 Formula Weight: 142.11 Melting Point: 127-132° MDL No.: MFCD00000625
Formel: C₅H₆N₂O₃
MW: 142,11 g/mol
Smältpunkt: 127…132 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002112
CAS nummer: 3849-21-6
EINECS: 223-351-3