Trimethyl-4-phosphonocrotonate mixture E/Z ≥90%
Leverantör: Thermo Scientific
Varning
A12399.09 A12399.18
A12399.09EA 2175 SEK
A12399.09 A12399.18
Trimethyl-4-phosphonocrotonate mixture E/Z ≥90%
Trimethyl-4-phosphonocrotonate
Purity: 90+% CAS No.: 86120-40-3 Molecular Formula: C7H13O5P Formula Weight: 208.15 Boiling Point: 123-124°/1mm Beilstein Reference: 4987133 MDL No.: MFCD00051564
Formel: C₇H₁₃O₅P
MW: 208,15 g/mol
Kokpunkt: 123…124 °C (1 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD00051564
CAS nummer: 86120-40-3