Cyclohexanone-2,4-dinitrophenylhydrazone 99%
Leverantör: Thermo Scientific
A12203.03 A12203.06 A12203.14
A12203.03EA 1225 SEK
A12203.03 A12203.06 A12203.14
Cyclohexanone-2,4-dinitrophenylhydrazone 99%
Cyclohexanone-2,4-dinitrophenylhydrazone
Purity: 99% CAS No.: 1589-62-4 Molecular Formula: C12H14N4O4 Formula Weight: 278.27 Melting Point: 158-160° MDL No.: MFCD00001658
Formel: C₁₂H₁₄N₄O₄
MW: 278,27 g/mol
Smältpunkt: 158…160 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001658
CAS nummer: 1589-62-4
EINECS: 216-458-1