1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid 97%
Leverantör: Thermo Scientific
Synonyms: 1-Ethyl-3-methylpyrazole-5-carboxylic acid
A11561.03 A11561.06
A11561.03EA 1450 SEK
A11561.03 A11561.06
1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid 97%
1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
Purity: 97% CAS No.: 50920-65-5 Molecular Formula: C7H10N2O2 Formula Weight: 154.17 Melting Point: 137-140° MDL No.: MFCD00156147
Formel: C₇H₁₀N₂O₂
MW: 154,17 g/mol
Smältpunkt: 137…140 °C
Storage Temperature: Omgivande
MDL Number: MFCD00156147
CAS nummer: 50920-65-5