4-Methoxy-3-buten-2-one 90%, tech.
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Methoxybut-3-en-2-one
A11235.09 A11235.18 A11235.30
A11235.09EA 1250 SEK
A11235.09 A11235.18 A11235.30
4-Methoxy-3-buten-2-one 90%, tech.
4-Methoxy-3-buten-2-one
Purity: 90% CAS No.: 4652-27-1 Molecular Formula: C5H8O2 Formula Weight: 100.12 Storage Temperature: -30°C to -10°C Flash Point: 63°(145°F) Refractive Index: 1.4650 Boiling Point: 66-69°/12mm Beilstein Reference: 1741053 MDL No.: MFCD00008778
Formel: C₅H₈O₂
MW: 100,117 g/mol
Kokpunkt: 66…69 °C (12 mmHg)
Densitet: 0,990
Flampunkt: 63 °C (145 °F)
Storage Temperature: Frys
MDL Number: MFCD00008778
CAS nummer: 4652-27-1
EINECS: 225-087-4