Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers), tech.
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Methylnadic Anhydride, Methylnadic anhydride, Nadic Methyl Anhydride, Methyl-3,6-endomethylenetetrahydrophthalic Anhydride
A10982.36 A10982.0E
A10982.36EA 2600 SEK
A10982.36 A10982.0E
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers), tech.
Methyl-5-norbornene-2,3-dicarboxylic anhydride (mixture of isomers)
CAS No.: 25134-21-8 Molecular Formula: C10H10O3 Formula Weight: 178.19 Flash Point: 135°(275°F) Refractive Index: 1.5052 Boiling Point: 137-140°/10mm MDL No.: MFCD00167036
Formel: C₁₀H₁₀O₃
Kokpunkt: 137…140 °C (10 mmHg)
Densitet: 1,232
Flampunkt: 135 °C (275 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00167036
CAS nummer: 25134-21-8
EINECS: 246-644-8
UN: 2810
ADR: 6.1,III