DL-Tartaric acid 99%
Leverantör: Thermo Scientific
Varning
Synonyms: (2R,3R)-rel-2,3-Dihydroxysuccinic acid, Racemic Acid
A10683.0B A10683.30 A10683.36
A10683.0BEA 3625 SEK
A10683.0B A10683.30 A10683.36
DL-Tartaric acid 99%
DL-Tartaric acid
Purity: 99% CAS No.: 133-37-9 Molecular Formula: C4H6O6 Formula Weight: 150.09 Melting Point: 210-212° dec. Beilstein Reference: 1725148 MDL No.: MFCD00071626
Formel: C₄H₆O₆
MW: 150,09 g/mol
Smältpunkt: 210…212 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00151254
CAS nummer: 133-37-9
EINECS: 205-105-7
Merck Index: 14,9069