Cerium(IV)oxid 20% i vatten, nanoparticle dispersion <5.0nm APS Number Weighted, high pH
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Ceric oxide, Cerium dioxide
47240.22 47240.36
47240.22EA 4675 SEK
47240.22 47240.36
Cerium(IV)oxid 20% i vatten, nanoparticle dispersion <5.0nm APS Number Weighted, high pH
Cerium(IV)oxid
CAS No.: 1306-38-3 Molecular Formula: CeO2 Formula Weight: 172.12 Physical Form: Liquid Density: 1.23 Appearance: Red-brown Odour: Odourless pH: 8.0 MDL No.: MFCD00010933
Formel: CeO₂
MW: 172.11 g/mol
Kokpunkt: 3468
Smältpunkt: 2000
Densitet: 1.23
Storage Temperature: Omgivande
MDL Number: MFCD00010933
CAS nummer: 1306-38-3
EINECS: 215-150-4
Merck Index: 12,02035