Chromium(II) chloride ≥99.9% -80 mesh
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Chromium dichloride
47176.03 47176.06 47176.14
47176.03EA 1325 SEK
47176.03 47176.06 47176.14
Chromium(II) chloride ≥99.9% -80 mesh
Chromium(II) chloride
Purity: 99.9% CAS No.: 10049-05-5 Molecular Formula: Cl2Cr Formula Weight: 122.9 Physical Form: -80 Mesh Powder Density: 2.878 Sensitivity: Air and Moisture Sensitive Appearance: Grey Odour: Odourless Melting Point: 824° UN No.: UN3260 MDL No.: MFCD00010947
Formel: Cl₂Cr
MW: 122.9 g/mol
Kokpunkt: 1302
Smältpunkt: 824 °C
Densitet: 2.878
Storage Temperature: Omgivande
MDL Number: MFCD00010947
CAS nummer: 10049-05-5
EINECS: 233-163-3
UN: 3260
ADR: 8 III
Merck Index: 14,2244