Rubidium fluoride ≥99.1%
Leverantör: Thermo Scientific
|
Fara
|
47144.09
47144.18
47144.09EA
1475
SEK
47144.09
47144.18
Rubidium fluoride ≥99.1%
Rubidium fluoride
Purity: 99.1%
CAS No.: 13446-74-7
Molecular Formula: FRb
Formula Weight: 104.47
Physical Form: Crystalline
Density: 3.557
Refractive Index: 1.398
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 795°
UN No.: UN3288
MDL No.: MFCD00011191
Formel:
FRb MW: 104,47 g/mol Smältpunkt: 795 °C Densitet: 3,557 Storage Temperature: Omgivande |
MDL Number:
MFCD00011191 CAS nummer: 13446-74-7 EINECS: 236-603-2 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...