Kaliumbromid, Premium FTIR Grade
Leverantör: Thermo Scientific
Varning
47111.KD 47111.KM 47111.KN 47111.KQ
47111.KDEA 4800 SEK
47111.KD 47111.KM 47111.KN 47111.KQ
Kaliumbromid, Premium FTIR Grade
Potassium bromide
CAS No.: 7758-02-3 Molecular Formula: BrK Formula Weight: 119.01 Physical Form: Powder in 0.5g Packets Density: 2.75 Refractive Index: 1.559 Odour: Odourless Melting Point: 730° Boiling Point: 1435° MDL No.: MFCD00011358
Formel: KBr
MW: 119 g/mol
Kokpunkt: 1435 °C
Smältpunkt: 730 °C
Densitet: 2,75
Storage Temperature: Omgivande
MDL Number: MFCD00011358
CAS nummer: 7758-02-3
EINECS: 231-830-3
Merck Index: 14,7618