Strontium bis(trifluoromethylsulphonyl)imide
Leverantör: Thermo Scientific
Fara
Synonyms: Bis(trifluoromethane)sulphonimide strontium salt, Bis(trifluoromethanesulphonyl)imide strontium salt
47099.03 47099.06
47099.03EA 2950 SEK
47099.03 47099.06
Strontium bis(trifluoromethylsulphonyl)imide
Strontium bis(trifluoromethylsulphonyl)imide
CAS No.: 862121-57-1 Molecular Formula: C4F12N2O8S4Sr Formula Weight: 647.91 Physical Form: Solid Sensitivity: Hygroscopic Appearance: Off-white Melting Point: 200° UN No.: UN3261 MDL No.: MFCD23380176
Formel: C₄F₁₂N₂O₈S₄Sr
MW: 647,91 g/mol
Smältpunkt: >200 °C
Storage Temperature: Omgivande
MDL Number: MFCD23380176
CAS nummer: 862121-57-1
ADR: 8,III