Cobalt bis(trifluoromethylsulphonyl)imide
Leverantör: Thermo Scientific
Fara
Synonyms: Bis(trifluoromethanesulphonyl)imide cobalt salt, Bis(trifluoromethane)sulphonimide cobalt salt
47053.03 47053.06
47053.03EA 3050 SEK
47053.03 47053.06
Cobalt bis(trifluoromethylsulphonyl)imide
Cobalt bis(trifluoromethylsulphonyl)imide
CAS No.: 207861-61-8 Molecular Formula: C4CoF12N2O8S4 Formula Weight: 619.23 Physical Form: Solid Sensitivity: Hygroscopic Appearance: Pale purple to purple Melting Point: 200° UN No.: UN3261 MDL No.: MFCD23380169
Formel: C₄CoF₁₂N₂O₈S₄
MW: 619,23 g/mol
Smältpunkt: >200 °C
Storage Temperature: Omgivande
MDL Number: MFCD23380169
CAS nummer: 207861-61-8
ADR: 8,III