Palladium 10% på aktivt kol wetted with ca. 50% water, pulver Type A402023-10
Leverantör: Thermo Scientific
46789.04 46789.09 46789.18
46789.04EA 1600 SEK
46789.04 46789.09 46789.18
Palladium 10% på aktivt kol wetted with ca. 50% water, pulver Type A402023-10
Palladium
CAS No.: 7440-05-3 Physical Form: Sold on a dry weight basis Appearance: Black MDL No.: MFCD03457879
Formel: Pd
MW: 106,420 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03457879
CAS nummer: 7440-05-3