Aluminiumoxid neutral HPLC flash grade
Leverantör: Thermo Scientific
46025.A1 46025.30
46025.A1EA 5675 SEK
46025.A1 46025.30
Aluminiumoxid neutral HPLC flash grade
Aluminium oxide neutral
CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 18-32 Micron APS Powder Refractive Index: 1.765 Appearance: White Odour: Odourless Melting Point: 2045° Boiling Point: 2980° MDL No.: MFCD00003424
Formel: Al₂O₃
MW: 101,96 g/mol
Kokpunkt: 2980 °C
Smältpunkt: 2045 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356