Hydroxipropylmetylcellulosa (Hypromellose), 7500-14000 mPa.s (2% in H₂O, 20 °C) 28-30% methoxyl, 7-12% hydroxypropyl
Leverantör: Thermo Scientific
Synonyms: Methocel K100, Hydroxypropyl methyl cellulose
45847.A4 45847.22 45847.36
45847.A4EA 13500 SEK
45847.A4 45847.22 45847.36
Hydroxipropylmetylcellulosa (Hypromellose), 7500-14000 mPa.s (2% in H₂O, 20 °C) 28-30% methoxyl, 7-12% hydroxypropyl
Hydroxypropylmethylcellulose
CAS No.: 9004-65-3 Physical Form: Powder MDL No.: MFCD00131360
Formel: C₃H₈O₂·xCH₄O
Storage Temperature: Omgivande
MDL Number: MFCD00131360
CAS nummer: 9004-65-3
Merck Index: 14,4842