Palladium hydroxide (5% Pd) på kol (Pearlman's catalyst) wetted with ca. 50% water, pulver Type A403002-5
Leverantör: Thermo Scientific
45576.04 45576.09
45576.04EA 1925 SEK
45576.04 45576.09
Palladium hydroxide (5% Pd) på kol (Pearlman's catalyst) wetted with ca. 50% water, pulver Type A403002-5
Palladium hydroxide
CAS No.: 12135-22-7 Appearance: Black Odour: Odourless MDL No.: MFCD00064599
Formel: H₂O₂Pd
MW: 140,435 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00064599
CAS nummer: 12135-22-7
EINECS: 235-219-2