5'-O-(4,4'-Dimethoxytrityl)thymidine ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: DMT-T, 1-((2R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxytetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, 5'-O-Dimethoxytrityl-deoxythymidine
44815.02 44815.04 44815.09
44815.02EA 3650 SEK
44815.02 44815.04 44815.09
5'-O-(4,4'-Dimethoxytrityl)thymidine ≥98%
5'-O-(4,4'-Dimethoxytrityl)thymidine
Purity: 98+% CAS No.: 40615-39-2 Molecular Formula: C31H32N2O7 Formula Weight: 544.60 Physical Form: Powder MDL No.: MFCD00010113
Formel: C₃₁H₃₂N₂O₇
MW: 544,60 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00010113
CAS nummer: 40615-39-2
EINECS: 255-003-1