Poly(3-octylthiophene-2,5-diyl) regiorandom low metals
Leverantör: Thermo Scientific
44778.01
44778.03
44778.01EA
4525
SEK
44778.01
44778.03
Poly(3-octylthiophene-2,5-diyl) regiorandom low metals
Poly(3-octylthiophene-2,5-diyl)
CAS No.: 104934-51-2
Physical Form: Powder
Solubility: Soluble in THF, CHCl3
MDL No.: MFCD00241412
Applications: Conducting polymers
Storage Temperature:
Omgivande |
MDL Number:
MFCD00241412 CAS nummer: 104934-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...