Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 139,000
Leverantör: Thermo Fisher Scientific
44644.18 44644.30 44644.A1
44644.18EA 800 SEK
44644.18 44644.30 44644.A1
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 139,000
Polydimethylsiloxane, trimethylsiloxy terminated
CAS No.: 9016-00-6 Molecular Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3 Flash Point: 321°(610°F) Physical Form: Viscous liquid Density: 0.977 Refractive Index: 1.4035 Appearance: Colourless Odour: Odourless Melting Point: -40° Boiling Point: 205° MDL No.: MFCD00148360
Formel: (CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
Kokpunkt: >205 °C
Smältpunkt: < –40 °C
Densitet: 0.977
Flampunkt: 321 °C (610 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00148360
CAS nummer: 9016-00-6