STYRENE atactic, M.W. 125,000-250,000
Leverantör: Thermo Fisher Scientific
44537.09 44537.18
44537.09EA 895 SEK
44537.09 44537.18
STYRENE atactic, M.W. 125,000-250,000
Poly(styrene)
CAS No.: 9003-53-6 Formula Weight: 125,000-250,000 Physical Form: Beads Odour: Odourless MDL No.: MFCD00084450
Formel: C₈H₈
MW: 125000 - 250000 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00084450
CAS nummer: 9003-53-6