Magnesiumhydroxidkarbonat tetrahydrate (∼40 - 43,5% MgO), Reagent Grade
Leverantör: Thermo Fisher Scientific
Synonyms: Magnesium carbonate basic, Magnesium carbonate hydroxide, Magnesium carbonate hydrated basic
33333.36 33333.22
33333.36EA 2050 SEK
33333.36 33333.22
Magnesiumhydroxidkarbonat tetrahydrate (∼40 - 43,5% MgO), Reagent Grade
Magnesiumhydroxidkarbonat tetrahydrate
CAS No.: 39409-82-0 Molecular Formula: 4MgCO3.Mg(OH)2.4H2O approx. Formula Weight: 467.67 (395.58anhy) Physical Form: -325 Mesh Powder Appearance: White Odour: Odourless Merck Reference: 14,5660 MDL No.: MFCD00149783
Formel: 4MgCO₃·Mg(OH)₂·4H₂O
MW: 467.67 g/mol (395.58 anhy)
Smältpunkt: > 2500 °C
Densitet: 2.16 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149783
CAS nummer: 235-192-7
EINECS: 235-192-7
Merck Index: 14,5660