Polyethylene 1000 micron, low density
Leverantör: Thermo Fisher Scientific
43949.18 43949.30 43949.A1
43949.18EA 900 SEK
43949.18 43949.30 43949.A1
Polyethylene 1000 micron, low density
Polyethylene (CAS 9002-88-4)
CAS No.: 9002-88-4 Molecular Formula: (CH2CH2)n Physical Form: Granular Odour: Odourless MDL No.: MFCD00084423
Formel: (CH₂CH₂)n
Storage Temperature: Omgivande
MDL Number: MFCD00084423
CAS nummer: 9002-88-4
Merck Index: 14,7567