Tropaeolin O 0,04% w/v aqueous solution
Leverantör: Thermo Fisher Scientific
Synonyms: Sodium 2,4-Dihydroxyazobenzene-4'-sulfonate, 2,4-Dihydroxyazobenzene-4'-sulfonic Acid Sodium Salt, Acid Orange 6, Resorcinol Yellow
42772.AE 42772.AP
42772.AEEA 820 SEK
42772.AE 42772.AP
Tropaeolin O 0,04% w/v aqueous solution
Tropaeolin O
Molecular Formula: C12H9N2NaO5S Formula Weight: 316.27 Physical Form: Liquid MDL No.: MFCD00007499
Formel: C₁₂H₉N₂O₅SNa
MW: 316.26 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00007499
CAS nummer: 547-57-9