Polyethylene ∼ 400 micron, low density
Leverantör: Thermo Fisher Scientific
42607.06 42607.14 42607.22 42607.0E 42607.36
42607.06EA 625 SEK
42607.06 42607.14 42607.22 42607.0E 42607.36
Polyethylene ∼ 400 micron, low density
Polyethylene (CAS 9002-88-4)
CAS No.: 9002-88-4 Molecular Formula: (CH2CH2)n Physical Form: Powder Density: 0.92 Odour: Odourless MDL No.: MFCD00084423
Formel: (CH₂CH₂)n
Densitet: 0,92
Storage Temperature: Omgivande
MDL Number: MFCD00084423
CAS nummer: 9002-88-4
Merck Index: 14,7567