Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 28,000
Leverantör: Thermo Scientific
42503.AD 42503.AK 42503.K2
42503.ADEA 745 SEK
42503.AD 42503.AK 42503.K2
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 28,000
Polydimethylsiloxane, trimethylsiloxy terminated
CAS No.: 9016-00-6 Molecular Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3 Flash Point: 315°(599°F) Physical Form: Viscous liquid Density: 0.971 Refractive Index: 1.4034 Appearance: Colourless Odour: Odourless Melting Point: -40° Boiling Point: 205° MDL No.: MFCD00148360
Formel: (CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
Kokpunkt: >205 °C
Smältpunkt: < –40 °C
Densitet: 0,971
Flampunkt: 315 °C (599 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00148360
CAS nummer: 9016-00-6