Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 2000
Leverantör: Thermo Scientific
42499.AD 42499.AK 42499.K2
42499.ADEA 835 SEK
42499.AD 42499.AK 42499.K2
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 2000
Polydimethylsiloxane, trimethylsiloxy terminated
CAS No.: 9016-00-6 Molecular Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3 Flash Point: 232°(450°F) Physical Form: Liquid Density: 0.966 Refractive Index: 1.4000 Appearance: Colourless Odour: Odourless Melting Point: -40° Boiling Point: 205° MDL No.: MFCD00148360
Formel: (CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
Kokpunkt: >205 °C
Smältpunkt: < –40 °C
Densitet: 0,966
Flampunkt: 232 °C (450 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00148360
CAS nummer: 9016-00-6